missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-L-leucine [S-(E)]-1-(2-Nonenyl)dodecyl Ester, TRC

Product Code. 30406788
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30406788 25 mg
30390239 5 mg
30397291 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406788 Supplier Toronto Research Chemicals Supplier No. TRCF70057525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130676-63-0
Molecular Formula C28 H53 N O3
Molecular Weight (g/mol) 451.73
Synonym N-Formyl-L-leucine [S-(E)]-1-(2-Nonenyl)dodecyl Ester (9Cl)
IUPAC Name [(E,10S)-henicos-7-en-10-yl] (2S)-2-formamido-4-methylpentanoate
SMILES CCCCCCCCCCC[C@@H](C\C=C\CCCCCC)OC(=O)[C@H](CC(C)C)NC=O

Specifications

InChI Formula InChI=1S/C28H53NO3/c1-5-7-9-11-13-14-16-18-20-22-26(21-19-17-15-12-10-8-6-2)32-28(31)27(29-24-30)23-25(3)4/h17,19,24-27H,5-16,18,20-23H2,1-4H3,(H,29,30)/b19-17+/t26-,27+/m1/s1
Quantity 25 mg
Formula Weight 451.4025
Shipping Condition Room Temperature
Chemical Name or Material N-Formyl-L-leucine [S-(E)]-1-(2-Nonenyl)dodecyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.