CAS RN 130676-63-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N-Formyl-L-leucine [S-(E)]-1-(2-Nonenyl)dodecyl Ester, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
N-Formyl-L-leucine [S-(E)]-1-(2-Nonenyl)dodecyl Ester, TRC
CAS: 130676-63-0 Molecular Formula: C28 H53 N O3 Molecular Weight (g/mol): 451.73 Synonym: N-Formyl-L-leucine [S-(E)]-1-(2-Nonenyl)dodecyl Ester (9Cl) IUPAC Name: [(E,10S)-henicos-7-en-10-yl] (2S)-2-formamido-4-methylpentanoate SMILES: CCCCCCCCCCC[C@@H](C\C=C\CCCCCC)OC(=O)[C@H](CC(C)C)NC=O