missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-DL-tryptophan, 98%, Thermo Scientific™

Product Code. 15400149
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15400149

Brand: Thermo Scientific Alfa Aesar H63446.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16108-03-5
Molecular Formula C12H12N2O3
Molecular Weight (g/mol) 232.24
MDL Number MFCD00055983
InChI Key RNEMLJPSSOJRHX-UHFFFAOYSA-N
Synonym n-formyl-dl-tryptophan, n-formyltryptophan, formyl-dl-trp-oh, nalpha-formyl-dl-tryptophan, 2-formamido-3-1h-indol-3-yl propanoic acid, 3-1h-indol-3-yl-2-formamidopropanoic acid, n-formyltryptophan #, tryptophan, n-formyl, for-dl-trp-oh, acmc-1clw6
PubChem CID 582651
IUPAC Name 2-formamido-3-(1H-indol-3-yl)propanoic acid
SMILES OC(=O)C(CC1=CNC2=CC=CC=C12)NC=O

Specifications

Quantity 250 mg
Formula Weight 232.24
Percent Purity 98%
Chemical Name or Material N-Formyl-DL-tryptophan
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.