CAS RN 16108-03-5
CAS RN 16108-03-5
IUPAC Name:
3-(1H-indol-3-yl)-2-formamidopropanoic acid
Synonyms:
3-(1H-indol-3-yl)-2-formamidopropanoic acid
Molecular Weight (g/mol):
232.24
Molecular Formula:
C12H12N2O3
InChi Key:
RNEMLJPSSOJRHX-UHFFFAOYNA-N
SMILES:
OC(=O)C(CC1=CNC2=CC=CC=C12)NC=O
1
–
1
of
1
results
1
N-Formyl-DL-tryptophan, 98%, Thermo Scientific™
CAS: 16108-03-5 Molecular Formula: C12H12N2O3 Molecular Weight (g/mol): 232.24 MDL Number: MFCD00055983 InChI Key: RNEMLJPSSOJRHX-UHFFFAOYSA-N Synonym: n-formyl-dl-tryptophan,n-formyltryptophan,formyl-dl-trp-oh,nalpha-formyl-dl-tryptophan,2-formamido-3-1h-indol-3-yl propanoic acid,3-1h-indol-3-yl-2-formamidopropanoic acid,n-formyltryptophan #,tryptophan, n-formyl,for-dl-trp-oh,acmc-1clw6 PubChem CID: 582651 IUPAC Name: 2-formamido-3-(1H-indol-3-yl)propanoic acid SMILES: OC(=O)C(CC1=CNC2=CC=CC=C12)NC=O