missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-leucine pentafluorophenyl ester, 98%, Thermo Scientific™

Product Code. 15460719
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15460719 1 g 1g
15470719 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15460719 Supplier Thermo Scientific Alfa Aesar Supplier No. H63820.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86060-88-0
Molecular Formula C27H22F5NO4
Molecular Weight (g/mol) 519.47
MDL Number MFCD00065658
InChI Key NTQJCLLWLHKJLU-TWYLJJHKNA-N
Synonym fmoc-leu-opfp, fmoc-l-leucine pentafluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-leucine pentafluorphenyl ester, 2,3,4,5,6-pentafluorophenyl 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylpentanoate, pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-l-leucinate, n-9h-fluorene-9-ylmethoxycarbonyl-l-leucine pentafluorophenyl ester, pentafluorophenyl n-9h-fluoren-9-yl methoxy carbonyl-l-leucinate
PubChem CID 13585944
IUPAC Name (2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoate
SMILES CC(C)C[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Color White
Beilstein 4731726
Quantity 1 g
Formula Weight 519.46
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Fmoc-L-leucine pentafluorophenyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.