accessibility menu, dialog, popup

UserName

CAS RN 86060-88-0

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methylpentanoate
Synonyms: 2,3,4,5,6-pentafluorophenyl 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-methylpentanoate
Molecular Weight (g/mol): 519.47
Molecular Formula: C27H22F5NO4
InChi Key: NTQJCLLWLHKJLU-UHFFFAOYNA-N
SMILES: CC(C)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results