missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-isoleucine, 98%, Thermo Scientific™

Product Code. p-7025591
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11494647

Brand: Thermo Scientific Alfa Aesar B21154.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Fmoc-L-isoleucine is an unnatural amino acid used for peptidomimetics. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in methanol (very faint turbidity).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71989-23-6
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD00037125
InChI Key QXVFEIPAZSXRGM-DJJJIMSYSA-N
Synonym fmoc-ile-oh, fmoc-l-isoleucine, n-fmoc-l-isoleucine, fmoc-l-ile-oh, n-9h-fluoren-9-ylmethoxy carbonyl-l-isoleucine, n-9-fluorenylmethoxycarbonyl-l-isoleucine, fmoc-ile, fmoc-l-ile, 2s,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylpentanoic acid, 9-fluorenylmethoxycarbonyl-l-isoleucine
PubChem CID 2724629
SMILES CC[C@H](C)[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 146°C to 148°C
Quantity 1 g
Beilstein 4716717
Solubility Information Soluble in methanol (very faint turbidity).
IUPAC Name (2S,3S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylpentanoic acid
Formula Weight 353.42
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-isoleucine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.