accessibility menu, dialog, popup

UserName

CAS RN 71989-23-6

H3C(S)CH3(S)NHOOOHO

CAS RN 71989-23-6

IUPAC Name: (2S,3R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylpentanoate
Synonyms: (2S,3R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-methylpentanoate
Molecular Weight (g/mol): 352.41
Molecular Formula: C21H22NO4
InChi Key: QXVFEIPAZSXRGM-YJYMSZOUSA-M
SMILES: CC[C@@H](C)[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)