missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-glutamic acid 5-allyl ester, 95%

Product Code. 15432739
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15432739

Brand: Thermo Scientific Alfa Aesar H66385.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 133464-46-7 | C23H23NO6 | 409.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133464-46-7
Molecular Formula C23H23NO6
Molecular Weight (g/mol) 409.44
MDL Number MFCD00190879
InChI Key LRBARFFNYOKIAX-UHFFFAOYNA-N
Synonym fmoc-glu oall-oh, fmoc-l-glutamic acid 5-allyl ester, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy-5-oxopentanoic acid, ambotzfaa1372, 5-allyl n-fmoc-l-glutamate, n-fmoc-l-glutamic acid 5-allyl ester, fmoc-glu oall-oh hplc, 5-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 5-allyl ester, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid 5-allyl ester
PubChem CID 7019067
SMILES OC(=O)C(CCC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Color White
Beilstein 6670846
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-prop-2-enoxypentanoic acid
Quantity 5 g
Formula Weight 409.44
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Fmoc-L-glutamic acid 5-allyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.