accessibility menu, dialog, popup

UserName

CAS RN 133464-46-7

H2COOOHONHOO

CAS RN 133464-46-7

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxo-5-(prop-2-en-1-yloxy)pentanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-oxo-5-(prop-2-en-1-yloxy)pentanoic acid
Molecular Weight (g/mol): 409.44
Molecular Formula: C23H23NO6
InChi Key: LRBARFFNYOKIAX-UHFFFAOYNA-N
SMILES: OC(=O)C(CCC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)