missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-homophenylalanine, 98%, Thermo Scientific™

Product Code. 15432819
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15432819 5 g 5g
15422819 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15432819 Supplier Thermo Scientific Alfa Aesar Supplier No. H66746.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135944-09-1
Molecular Formula C25H23NO4
Molecular Weight (g/mol) 401.462
MDL Number MFCD00151917
InChI Key CIHPCIUGLIZADU-HSZRJFAPSA-N
Synonym fmoc-d-homophe-oh, fmoc-d-homophenylalanine, fmoc-d-hophe-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-phenylbutanoic acid, fmoc-d-hphe-oh, fmoc-d-hph-oh, fmoc-d-homophenyl-ala, fmoc-d-homophe, fmoc-d-hfe-oh, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-phenylbutanoic acid
PubChem CID 7129818
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-phenylbutanoic acid
SMILES C1=CC=C(C=C1)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Color White
Beilstein 4847668
Quantity 5 g
Formula Weight 401.46
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Fmoc-D-homophenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.