CAS RN 135944-09-1
CAS RN 135944-09-1
IUPAC Name:
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-phenylbutanoic acid
Synonyms:
2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-phenylbutanoic acid
Molecular Weight (g/mol):
401.46
Molecular Formula:
C25H23NO4
InChi Key:
CIHPCIUGLIZADU-UHFFFAOYNA-N
SMILES:
OC(=O)C(CCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
N-Fmoc-D-homophenylalanine, 98%, Thermo Scientific™
CAS: 135944-09-1 Molecular Formula: C25H23NO4 Molecular Weight (g/mol): 401.462 MDL Number: MFCD00151917 InChI Key: CIHPCIUGLIZADU-HSZRJFAPSA-N Synonym: fmoc-d-homophe-oh,fmoc-d-homophenylalanine,fmoc-d-hophe-oh,r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-phenylbutanoic acid,fmoc-d-hphe-oh,fmoc-d-hph-oh,fmoc-d-homophenyl-ala,fmoc-d-homophe,fmoc-d-hfe-oh,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-phenylbutanoic acid PubChem CID: 7129818 IUPAC Name: (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-phenylbutanoic acid SMILES: C1=CC=C(C=C1)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24