missing translation for 'onlineSavingsMsg'
Learn More

N-Ethoxycarbonyl Norfloxacin, TRC

Product Code. 17904755
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17904755 25 mg 25mg
17953565 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17904755 Supplier Toronto Research Chemicals Supplier No. TRCE67827525MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105440-01-5
Molecular Formula C19 H22 F N3 O5
Molecular Weight (g/mol) 391.39
Synonym 7-[4-(Ethoxycarbonyl)piperazin-1-yl]-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid
IUPAC Name 7-(4-ethoxycarbonylpiperazin-1-yl)-1-ethyl-6-fluoro-4-oxoquinoline-3-carboxylic acid
SMILES CCOC(=O)N1CCN(CC1)c2cc3N(CC)C=C(C(=O)O)C(=O)c3cc2F

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H22FN3O5/c1-3-21-11-13(18(25)26)17(24)12-9-14(20)16(10-15(12)21)22-5-7-23(8-6-22)19(27)28-4-2/h9-11H,3-8H2,1-2H3,(H,25,26)
Formula Weight 391.1543
Chemical Name or Material 7-[4-(ethoxycarbonyl)piperazin-1-yl]-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.