CAS RN 105440-01-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Ethoxycarbonyl Norfloxacin, TRC
CAS: 105440-01-5 Molecular Formula: C19 H22 F N3 O5 Molecular Weight (g/mol): 391.39 Synonym: 7-[4-(Ethoxycarbonyl)piperazin-1-yl]-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid IUPAC Name: 7-(4-ethoxycarbonylpiperazin-1-yl)-1-ethyl-6-fluoro-4-oxoquinoline-3-carboxylic acid SMILES: CCOC(=O)N1CCN(CC1)c2cc3N(CC)C=C(C(=O)O)C(=O)c3cc2F