missing translation for 'onlineSavingsMsg'
Learn More

N-Des-1-Beta-D-ribofuranosyl Ribavirin, TRC

Product Code. 17951865
Change view
Click to view available options
Quantity:
2.5 g
10 g
25 g
Unit Size:
10g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17951865 10 g 10g
17931555 25 g 25g
17922415 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17951865 Supplier Toronto Research Chemicals Supplier No. TRCD29736510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3641-08-5
Molecular Formula C3 H4 N4 O
Molecular Weight (g/mol) 112.09
Synonym 1H-1,2,4-Triazole-3-carboxamide, Ribavirin Imp. D (EP)
IUPAC Name 1H-1,2,4-triazole-3-carboxamide
SMILES NC(=O)c1nc[nH]n1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C3H4N4O/c4-2(8)3-5-1-6-7-3/h1H,(H2,4,8)(H,5,6,7)
Formula Weight 112.0385
Chemical Name or Material 1H-1,2,4-Triazole-3-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.