CAS RN 3641-08-5
CAS RN 3641-08-5
IUPAC Name:
1H-1,2,4-triazole-5-carboxamide
Synonyms:
2H-1,2,4-triazole-3-carboxamide
Molecular Weight (g/mol):
112.09
Molecular Formula:
C3H4N4O
InChi Key:
ZEWJFUNFEABPGL-UHFFFAOYSA-N
SMILES:
NC(=O)C1=NC=NN1
1
–
1
of
1
results
1
N-Des-1-Beta-D-ribofuranosyl Ribavirin, TRC
CAS: 3641-08-5 Molecular Formula: C3 H4 N4 O Molecular Weight (g/mol): 112.09 Synonym: 1H-1,2,4-Triazole-3-carboxamide,Ribavirin Imp. D (EP) IUPAC Name: 1H-1,2,4-triazole-3-carboxamide SMILES: NC(=O)c1nc[nH]n1