missing translation for 'onlineSavingsMsg'
Learn More

n-Butylurea, 96%

Product Code. 11337875
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337875 500 g 500g
11327875 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337875 Supplier Thermo Scientific Alfa Aesar Supplier No. L02854.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 592-31-4 | C5H12N2O | 116.164 g/mol

n-Butylurea is used as a fertilizer for rice. It is used in the preparation of drugs which are recommended for diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Butylurea is used as a fertilizer for rice. It is used in the preparation of drugs which are recommended for diabetes.

Solubility
Soluble in water, Alcohol, Ether.

Notes
Incompatible with Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-31-4
Molecular Formula C5H12N2O
Molecular Weight (g/mol) 116.164
MDL Number MFCD00007956
InChI Key CNWSQCLBDWYLAN-UHFFFAOYSA-N
Synonym n-butylurea, 1-butylurea, urea, butyl, urea, n-butyl, unii-9cpl5nr15k, n-butyl urea, 9cpl5nr15k, dsstox_cid_222, dsstox_rid_75443, dsstox_gsid_20222
PubChem CID 11595
IUPAC Name butylurea
SMILES CCCCNC(=O)N

Specifications

Melting Point 92°C to 96°C
Quantity 500 g
Beilstein 1744775
Solubility Information Soluble in water,Alcohol,Ether.
Formula Weight 116.16
Percent Purity 96%
Chemical Name or Material n-Butylurea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.