accessibility menu, dialog, popup

UserName

H3CHNNH2O

CAS RN 592-31-4

IUPAC Name: butylurea
Synonyms: butylurea
Molecular Weight (g/mol): 116.16
Molecular Formula: C5H12N2O
InChi Key: CNWSQCLBDWYLAN-UHFFFAOYSA-N
SMILES: CCCCNC(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results