missing translation for 'onlineSavingsMsg'
Learn More

n-Butylmagnesium chloride, 1.5-2.5M in THF, Thermo Scientific™

Product Code. 15462617
Change view
Click to view available options
Quantity:
50 mL
250 mL
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15462617 50 mL 50mL
15472617 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15462617 Supplier Thermo Scientific Alfa Aesar Supplier No. 046117.AD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-04-9
Molecular Formula C4H9ClMg
Molecular Weight (g/mol) 116.87
MDL Number MFCD00000475
InChI Key UUNGXYVLXYGZKH-UHFFFAOYSA-M
Synonym butylmagnesium chloride, magnesium, butylchloro, nbumgcl, bumgcl, butylmagnesiumchloride, n-butylmagnesium chloride, n-butyl-magnesium chloride, butylmagnesium chloride solution, butylmagnesium chloride solution, 2.0 m in diethyl ether, unii-4j83fgw852
PubChem CID 101837
IUPAC Name magnesium;butane;chloride
SMILES CCCC[Mg]Cl

Specifications

Color Brown
Odor Ether-like
Linear Formula CH3(CH2)3MgCl
Quantity 50 mL
UN Number UN3399
Beilstein 3587228
Sensitivity Air and moisture sensitive
Formula Weight 116.87
Concentration or Composition (by Analyte or Components) 1.5 to 2.5M in THF
Physical Form Liquid
Chemical Name or Material n-Butylmagnesium chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.