accessibility menu, dialog, popup

UserName

H3CMgCl

CAS RN 693-04-9

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 116.87
Molecular Formula: C4H9ClMg
InChi Key: QUXHCILOWRXCEO-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].CCC[CH2-]
Physical Form 
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)