missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Butylmagnesium chloride, 1.5-2.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Packaged under Argon
Specifications
Specifications
| Color | Brown |
| Packaging | ChemSeal™ bottles |
| Linear Formula | CH3(CH2)3MgCl |
| Quantity | 50 mL |
| UN Number | UN3399 |
| Beilstein | 3587228 |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Reacts with water. |
| Formula Weight | 116.87 |
| Concentration or Composition (by Analyte or Components) | 1.5 to 2.5M in THF |
| Show More |
Safety and Handling
Product Identifier
- n-Butylmagnesium chloride, 1.5-2.5M in THF
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- Substance which, in contact with water, emits flammable gases Category 2
- H226-Flammable liquid and vapour.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H336-May cause drowsiness or dizziness.
- H360-May damage fertility or the unborn child.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Tetrahydrofuran, Butylmagnesium chloride
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?