missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Visocolor™ ECO Phosphate Colorimetric Test Kit

Product Code. 10161472 Shop All Macherey-Nagel Products
Change view
Click to view available options
For Use With: Colorimetric determination of phosphate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10161472 For Use With: Colorimetric determination of phosphate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10161472 Supplier Macherey-Nagel™ Supplier No. 931284
  • ONLINE EXCLUSIVE PRICE
    £31.33
    £28.16 / 50mL
    Save £3.17
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

For Use With (Application) Colorimetric Determination of Phosphate
compliance-icons compliance-icons
Product Identifier
  • visocolor eco phosphate, refill pack
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: ammonium heptamolybdate, sulphuric acid,sodium disulphite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.