missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-(methylamino)acetaldehyde, 97%

Product Code. 11489550
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11489550

Brand: Thermo Scientific Alfa Aesar H50870.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123387-72-4 | C8H15NO3 | 173.212 g/mol

N-Boc-(methylamino)acetaldehyde is involved in the preparation of benzo[d]imidazole inhibitor of co-activator associated arginine methyltransferase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-(methylamino)acetaldehyde is involved in the preparation of benzo[d]imidazole inhibitor of co-activator associated arginine methyltransferase.

Solubility
Slightly miscible with water.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123387-72-4
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.212
MDL Number MFCD08064267
InChI Key MSWTVSDFEYSRMQ-UHFFFAOYSA-N
Synonym n-boc-methylamino acetaldehyde, tert-butyl methyl 2-oxoethyl carbamate, tert-butyl n-methyl-n-2-oxoethyl carbamate, n-tert-butoxycarbonyl-n-methylaminoacetaldehyde, methyl-2-oxo-ethyl-carbamic acid tert-butyl ester, carbamic acid, methyl 2-oxoethyl-, 1,1-dimethylethyl ester, carbamic acid,n-methyl-n-2-oxoethyl-, 1,1-dimethylethyl ester, acmc-20a2gd, n-t-butoxycarbonylsarcosinal, n-boc-n-methyl-2-amino-acetaldehyde
PubChem CID 10725881
IUPAC Name tert-butyl N-methyl-N-(2-oxoethyl)carbamate
SMILES CC(C)(C)OC(=O)N(C)CC=O

Specifications

Solubility Information Slightly miscible with water.
Quantity 1 g
Formula Weight 173.21
Sensitivity Air sensitive
Percent Purity 98%
Chemical Name or Material N-Boc-(methylamino)acetaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.