accessibility menu, dialog, popup

UserName

CAS RN 123387-72-4

CH3CH3H3COONCH3O

CAS RN 123387-72-4

IUPAC Name: tert-butyl N-methyl-N-(2-oxoethyl)carbamate
Synonyms: tert-butyl N-methyl-N-(2-oxoethyl)carbamate
Molecular Weight (g/mol): 173.21
Molecular Formula: C8H15NO3
InChi Key: MSWTVSDFEYSRMQ-UHFFFAOYSA-N
SMILES: CN(CC=O)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)