Learn More
N-Boc-(methylamino)acetaldehyde, 97%
Description
N-Boc-(methylamino)acetaldehyde is involved in the preparation of benzo[d]imidazole inhibitor of co-activator associated arginine methyltransferase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-Boc-(methylamino)acetaldehyde is involved in the preparation of benzo[d]imidazole inhibitor of co-activator associated arginine methyltransferase.
Solubility
Slightly miscible with water.
Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Solubility Information | Slightly miscible with water. |
| Quantity | 250 mg |
| Formula Weight | 173.21 |
| Sensitivity | Air sensitive |
| Percent Purity | 98% |
| Chemical Name or Material | N-Boc-(methylamino)acetaldehyde |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.