missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-4-(Fmoc-amino)-L-phenylalanine, 95%, Thermo Scientific Chemicals
£43.90 - £120.00
Chemical Identifiers
CAS | 114346-31-5 |
---|---|
Molecular Formula | C29H30N2O6 |
Molecular Weight (g/mol) | 502.567 |
MDL Number | MFCD00151889 |
InChI Key | ZKSJJSOHPQQZHC-VWLOTQADSA-N |
Synonym | boc-4-fmoc-amino-l-phenylalanine, boc-p-amino-phe fmoc-oh, boc-l-4-fmoc aminophenylalanine, boc-phe 4-nhfmoc-oh, boc-l-4-aminophenylalanine fmoc, s-3-4-9h-fluoren-9-yl methoxy carbonyl amino phenyl-2-tert-butoxycarbonyl amino propanoic acid, s-boc-4-fmoc-amino-phenylalanine, boc-d-4-fmoc aminophenylalanine, n-1,1-dimethylethoxy carbonyl-4-9h-fluoren-9-ylmethoxy carbonyl amino-l-phenylalanine, n-alpha-t-butyloxycarbonyl-4-9-fluorenylmethyloxycarbonyl amino-l-phenylalanine |
PubChem CID | 2755959 |
IUPAC Name | (2S)-3-[4-(9H-fluoren-9-ylmethoxycarbonylamino)phenyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
SMILES | CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15410289
|
Thermo Scientific Alfa Aesar
H63543.03 |
1 g |
£43.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15420289
|
Thermo Scientific Alfa Aesar
H63543.06 |
5 g |
£120.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
114346-31-5 | |
502.567 | |
ZKSJJSOHPQQZHC-VWLOTQADSA-N | |
2755959 | |
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O |
C29H30N2O6 | |
MFCD00151889 | |
boc-4-fmoc-amino-l-phenylalanine, boc-p-amino-phe fmoc-oh, boc-l-4-fmoc aminophenylalanine, boc-phe 4-nhfmoc-oh, boc-l-4-aminophenylalanine fmoc, s-3-4-9h-fluoren-9-yl methoxy carbonyl amino phenyl-2-tert-butoxycarbonyl amino propanoic acid, s-boc-4-fmoc-amino-phenylalanine, boc-d-4-fmoc aminophenylalanine, n-1,1-dimethylethoxy carbonyl-4-9h-fluoren-9-ylmethoxy carbonyl amino-l-phenylalanine, n-alpha-t-butyloxycarbonyl-4-9-fluorenylmethyloxycarbonyl amino-l-phenylalanine | |
(2S)-3-[4-(9H-fluoren-9-ylmethoxycarbonylamino)phenyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
114346-31-5 | |
MFCD00151889 | |
ZKSJJSOHPQQZHC-VWLOTQADSA-N | |
(2S)-3-[4-(9H-fluoren-9-ylmethoxycarbonylamino)phenyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
2755959 | |
95% | |
N-Boc-4-(Fmoc-amino)-L-phenylalanine |
C29H30N2O6 | |
boc-4-fmoc-amino-l-phenylalanine, boc-p-amino-phe fmoc-oh, boc-l-4-fmoc aminophenylalanine, boc-phe 4-nhfmoc-oh, boc-l-4-aminophenylalanine fmoc, s-3-4-9h-fluoren-9-yl methoxy carbonyl amino phenyl-2-tert-butoxycarbonyl amino propanoic acid, s-boc-4-fmoc-amino-phenylalanine, boc-d-4-fmoc aminophenylalanine, n-1,1-dimethylethoxy carbonyl-4-9h-fluoren-9-ylmethoxy carbonyl amino-l-phenylalanine, n-alpha-t-butyloxycarbonyl-4-9-fluorenylmethyloxycarbonyl amino-l-phenylalanine | |
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O | |
502.567 | |
502.56 | |
Solid |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only