missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-(Fmoc-amino)-L-phenylalanine, 95%

Product Code. 15410289
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15410289

Brand: Thermo Scientific Alfa Aesar H63543.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 114346-31-5 | C29H30N2O6 | 502.567 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114346-31-5
Molecular Formula C29H30N2O6
Molecular Weight (g/mol) 502.567
MDL Number MFCD00151889
InChI Key ZKSJJSOHPQQZHC-VWLOTQADSA-N
Synonym boc-4-fmoc-amino-l-phenylalanine, boc-p-amino-phe fmoc-oh, boc-l-4-fmoc aminophenylalanine, boc-phe 4-nhfmoc-oh, boc-l-4-aminophenylalanine fmoc, s-3-4-9h-fluoren-9-yl methoxy carbonyl amino phenyl-2-tert-butoxycarbonyl amino propanoic acid, s-boc-4-fmoc-amino-phenylalanine, boc-d-4-fmoc aminophenylalanine, n-1,1-dimethylethoxy carbonyl-4-9h-fluoren-9-ylmethoxy carbonyl amino-l-phenylalanine, n-alpha-t-butyloxycarbonyl-4-9-fluorenylmethyloxycarbonyl amino-l-phenylalanine
PubChem CID 2755959
IUPAC Name (2S)-3-[4-(9H-fluoren-9-ylmethoxycarbonylamino)phenyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O

Specifications

Quantity 1 g
Formula Weight 502.56
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-4-(Fmoc-amino)-L-phenylalanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.