missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-cyclopentyl-L-glycine dicyclohexylammonium salt, 95%, Thermo Scientific™

Product Code. p-7054545
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
This item is not returnable. View return policy

Product Code. 15497896

Brand: Thermo Scientific Alfa Aesar H62838.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 676559-50-5
MDL Number MFCD05663437
Synonym boc-l-cyclopentylglycine, s-2-tert-butoxycarbonyl amino-2-cyclopentylacetic acid, s-tert-butoxycarbonylamino-cyclopentyl-acetic acid, s-tert-butoxycarbonyl amino cyclopentyl acetic acid, 2s-tert-butoxycarbonyl amino cyclopentyl ethanoic acid, 2s-2-tert-butoxy carbonyl amino-2-cyclopentylacetic acid, ambotzbaa5140, boc-l-cyclopentyl glycine, n-tert-butoxycarbonyl-l-cyclopentylglycine, n-alpha-t-butyloxycarbonyl-l-cyclopentylglycine
PubChem CID 7020746
IUPAC Name (2S)-2-cyclopentyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid

Specifications

Color White
Molecular Formula C24H44N2O4
InChI Key MCIPNICZJUEQHT-UHFFFAOYNA-N
SMILES C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NC(C1CCCC1)C(O)=O
Molecular Weight (g/mol) 424.63
Quantity 250 mg
Formula Weight 424.62
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-2-cyclopentyl-L-glycine dicyclohexylammonium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.