accessibility menu, dialog, popup

UserName

CAS RN 676559-50-5

CH3H3CCH3OOHNOOHHN

CAS RN 676559-50-5

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-2-cyclopentylacetic acid; N-cyclohexylcyclohexanamine
Synonyms: [(tert-butoxycarbonyl)amino](cyclopentyl)acetic acid; dicha
Molecular Weight (g/mol): 424.63
Molecular Formula: C24H44N2O4
InChi Key: MCIPNICZJUEQHT-UHFFFAOYNA-N
SMILES: C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NC(C1CCCC1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)