missing translation for 'onlineSavingsMsg'
Learn More

N-(Benzyloxycarbonyl)sarcosine, 95%

Product Code. 15478708
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15478708

Brand: Thermo Scientific Alfa Aesar H62135.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39608-31-6 | C11H13NO4 | 223.228 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39608-31-6
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.228
MDL Number MFCD00021747
InChI Key CBWFTZNMONHKNZ-UHFFFAOYSA-N
Synonym z-sar-oh, cbz-sar-oh, n-benzyloxycarbonyl sarcosine, carbobenzoxy sarcosine, z-sarcosine, glycine, n-methyl-n-phenylmethoxy carbonyl, n-cbz-sarcosine, 2-benzyloxy carbonyl methyl amino acetic acid, n-carbobenzoxy sarcosine, benzyloxycarbonyl sarcosine
PubChem CID 97053
IUPAC Name 2-[methyl(phenylmethoxycarbonyl)amino]acetic acid
SMILES CN(CC(=O)O)C(=O)OCC1=CC=CC=C1

Specifications

Quantity 5 g
Formula Weight 223.23
Percent Purity 95%
Chemical Name or Material N-(Benzyloxycarbonyl)sarcosine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.