accessibility menu, dialog, popup

UserName

CAS RN 39608-31-6

CH3NHOOOO

CAS RN 39608-31-6

IUPAC Name: 2-{[(benzyloxy)carbonyl](methyl)amino}acetic acid
Synonyms: {[(benzyloxy)carbonyl](methyl)amino}acetic acid
Molecular Weight (g/mol): 223.23
Molecular Formula: C11H13NO4
InChi Key: CBWFTZNMONHKNZ-UHFFFAOYSA-N
SMILES: CN(CC(O)=O)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)