missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Benzyl-N-(3-pyridylmethylene)amine

Product Code. 11444680
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11444680

Brand: Alfa Aesar™ H50745.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 71718-88-2
Molecular Formula C13H12N2
Molecular Weight (g/mol) 196.253
MDL Number MFCD08064239
InChI Key DXDPRQLAIWIMDP-UHFFFAOYSA-N
Synonym 1-phenyl-n-pyridin-3-ylmethylene methanamine, n-benzyl-n-3-pyridylmethylene amine, benzyl pyridin-3-ylmethylidene amine, acmc-1bded, n-benzylpyridine-3-methaneimine, benzylpyridin-3-ylmethylene-amine, e-n-benzyl-1-pyridin-3-yl methanimine, benzenemethanamine,n-3-pyridinylmethylene, e-benzyl pyridin-3-ylmethylidene amine, 1e-3-phenyl-1-3-pyridyl-2-azaprop-1-ene
PubChem CID 3851439
IUPAC Name N-benzyl-1-pyridin-3-ylmethanimine
SMILES C1=CC=C(C=C1)CN=CC2=CN=CC=C2

Specifications

Quantity 1 g
Formula Weight 196.25
Chemical Name or Material N-Benzyl-N-(3-pyridylmethylene)amine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.