accessibility menu, dialog, popup

UserName

CAS RN 71718-88-2

NN(E/Z)

CAS RN 71718-88-2

IUPAC Name: benzyl[(pyridin-3-yl)methylidene]amine
Synonyms: benzyl(pyridin-3-ylmethylidene)amine
Molecular Weight (g/mol): 196.25
Molecular Formula: C13H12N2
InChi Key: DXDPRQLAIWIMDP-UHFFFAOYSA-N
SMILES: C(N=CC1=CN=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results