missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ N-Benzyl-N-(3-pyridylmethylene)amine
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
missing translation for 'unitSize'
1g
250mg
Specifications
Specifications
| Quantity | 250 mg |
| Formula Weight | 196.25 |
| Chemical Name or Material | N-Benzyl-N-(3-pyridylmethylene)amine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction