missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetylglycine, 99%

Product Code. 11424957
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424957 500 g 500g
11414957 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424957 Supplier Thermo Scientific Chemicals Supplier No. B21887.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 543-24-8 | C4H7NO3 | 117.104 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-24-8
Molecular Formula C4H7NO3
Molecular Weight (g/mol) 117.104
MDL Number MFCD00004275
InChI Key OKJIRPAQVSHGFK-UHFFFAOYSA-N
Synonym n-acetylglycine, aceturic acid, acetamidoacetic acid, acetylglycine, ac-gly-oh, glycine, n-acetyl, acetylaminoacetic acid, acetylamino-acetic acid, acetylglycocoll, ethanoylaminoethanoic acid
PubChem CID 10972
ChEBI CHEBI:40410
IUPAC Name 2-acetamidoacetic acid
SMILES CC(=O)NCC(=O)O

Specifications

Melting Point ∼210°C (decomposition)
Beilstein 774114
Merck Index 14,80
Quantity 500 g
Formula Weight 117.1
Percent Purity 99%
Chemical Name or Material N-Acetylglycine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.