accessibility menu, dialog, popup

UserName

H3CONHOHO

CAS RN 543-24-8

IUPAC Name: 2-acetamidoacetic acid
Synonyms: acetylamino-acetic acid 15N-acetylglycine a-radical aceturic acid acetylglycocoll ac gly Ac-Gly-OH 醋甘氨酸 aceturate
Molecular Weight (g/mol): 117.1
Molecular Formula: C4H7NO3
InChi Key: OKJIRPAQVSHGFK-UHFFFAOYSA-N
SMILES: CC(=O)NCC(O)=O
Molecular Weight (g/mol) 
  • (7)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (5)
Physical Form 
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (7)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (5)
  • (3)
  • (1)