missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl Sulfamethazine, TRC

Product Code. 30132814
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132814 25 mg 25mg
17913535 5 mg 5mg
17914875 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132814 Supplier Toronto Research Chemicals Supplier No. TRCA18785025MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-90-3
Molecular Formula C14 H16 N4 O3 S
Molecular Weight (g/mol) 320.37
Synonym N-[4-[[(4,6-Dimethyl-2-pyrimidinyl)amino]sulfonyl]phenyl]acetamide, Acetanilide, 4'-[(4,6-dimethyl-2-pyrimidinyl)sulfamoyl]- (6CI,7CI,8CI), Acetanilide, p-(4,6-dimethyl-2-pyrimidylsulfamyl)- (4CI), 4'-[(4,6-Dimethyl-2-pyrimidinyl)sulfamoyl]acetanilide, Acetylsulfadimidine, Acetylsulfamethazine, N-Acetylsulfamethazine, N-[4-[[(4,6-Dimethyl-2-pyrimidinyl)amino]sulfonyl]phenyl]acetamide, N4-Acetylsulfadimidine, N4-Acetylsulfamethazine, N4-Acetylsulmet, Sulfamethazine-N4-acetyl
IUPAC Name N-[4-[(4,6-dimethylpyrimidin-2-yl)sulfamoyl]phenyl]acetamide
SMILES CC(=O)Nc1ccc(cc1)S(=O)(=O)Nc2nc(C)cc(C)n2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H16N4O3S/c1-9-8-10(2)16-14(15-9)18-22(20,21)13-6-4-12(5-7-13)17-11(3)19/h4-8H,1-3H3,(H,17,19)(H,15,16,18)
Formula Weight 320.0943
Percent Purity >95%
Chemical Name or Material N-Acetyl Sulfamethazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.