missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-DL-norvaline, 98%, Thermo Scientific™

Product Code. 11497066
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497066 1 g 1g
11407076 5 g 5g
11417076 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497066 Supplier Thermo Scientific Alfa Aesar Supplier No. B25389.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7682-15-7
Molecular Formula C7H13NO3
Molecular Weight (g/mol) 159.19
MDL Number MFCD00037269
InChI Key BSYFPUSAWVWWDG-UHFFFAOYSA-N
Synonym n-acetyl-dl-norvaline, ac-dl-nva-oh, acetyl-dl-norvaline, n-acetylnorvaline #, 2-acetamido-pentanoic acid, 2-acetylamino valeric acid, n-alpha-acetyl-dl-norvaline, n-acetyl-2-aminovaleric acid, 2-acetylamino pentanoic acid
PubChem CID 306106
IUPAC Name 2-acetamidopentanoic acid
SMILES CCCC(NC(C)=O)C(O)=O

Specifications

Quantity 1 g
Formula Weight 159.19
Percent Purity 98%
Chemical Name or Material N-Acetyl-DL-norvaline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.