accessibility menu, dialog, popup

UserName

CAS RN 7682-15-7

IUPAC Name: (2S)-2-acetamidopentanoate
Synonyms: (2S)-2-acetamidopentanoate
Molecular Weight (g/mol): 158.18
Molecular Formula: C7H12NO3
InChi Key: BSYFPUSAWVWWDG-LURJTMIESA-M
SMILES: CCC[C@H](NC(C)=O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results