missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide

Product Code. 15417119
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15417119 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15417119 Supplier Thermo Scientific™ Supplier No. J65878.MB
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 189950-66-1 | C26H34N6O13, C26H34N6O13 | 638.59 g/mol

N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide is an chromogenic substrate for Caspase 3 and CCP-32.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) A colorimetric substrate for caspase-3
CAS 189950-66-1
Molecular Formula C26H34N6O13
Formula Weight 638.58g/mol
Synonym Ac-DEVD-pNA
Product Type Signal Transduction Reagent
Form Powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.