missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-D-tryptophan, TRC

Product Code. 30389442
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389442 250 mg -
30389114 50 mg -
30391493 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389442 Supplier Toronto Research Chemicals Supplier No. TRCF547355250MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070774-51-4
Molecular Formula C27 H24 N2 O4
Molecular Weight (g/mol) 440.49
Synonym N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-D-tryptophan
IUPAC Name (2R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-(1H-indol-3-yl)propanoic acid
SMILES CN([C@H](Cc1c[nH]c2ccccc12)C(=O)O)C(=O)OCC3c4ccccc4c5ccccc35

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C27H24N2O4/c1-29(25(26(30)31)14-17-15-28-24-13-7-6-8-18(17)24)27(32)33-16-23-21-11-4-2-9-19(21)20-10-3-5-12-22(20)23/h2-13,15,23,25,28H,14,16H2,1H3,(H,30,31)/t25-/m1/s1
Formula Weight 440.1736
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-D-tryptophan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.