missing translation for 'onlineSavingsMsg'
Learn More

N,9-O-Diacetylneuraminic Acid, TRC

Product Code. 30383688
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383688 25 mg
30384644 10 mg
30377276 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383688 Supplier Toronto Research Chemicals Supplier No. TRCD45448325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55717-54-9
Molecular Formula C13H21NO10
Molecular Weight (g/mol) 351.31
Synonym 9-O-Acetyl-N-acetylneuraminic Acid, N-Acetyl-9-O-acetylneuraminic Acid, 9-Acetate-5-(acetylamino)-3,5-dideoxy-D-glycero-D-galacto-2-nonulosonic Acid
SMILES CC(=O)N[C@H]([C@@H](O)CC(=O)C(=O)O)[C@@H](O)[C@H](O)[C@H](O)COC(=O)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C13H21NO10/c1-5(15)14-9-7(17)3-13(22,12(20)21)24-11(9)10(19)8(18)4-23-6(2)16/h7-11,17-19,22H,3-4H2,1-2H3,(H,14,15)(H,20,21)/t7-,8+,9+,10+,11+,13-/m0/s1
Formula Weight 351.1165
Chemical Name or Material N,9-O-Diacetylneuraminic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.