CAS RN 55717-54-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N,9-O-Diacetylneuraminic Acid, TRC
CAS: 55717-54-9 Molecular Formula: C13H21NO10 Molecular Weight (g/mol): 351.31 Synonym: 9-O-Acetyl-N-acetylneuraminic Acid,N-Acetyl-9-O-acetylneuraminic Acid,9-Acetate-5-(acetylamino)-3,5-dideoxy-D-glycero-D-galacto-2-nonulosonic Acid SMILES: CC(=O)N[C@H]([C@@H](O)CC(=O)C(=O)O)[C@@H](O)[C@H](O)[C@H](O)COC(=O)C