| CAS |
169275-84-7
|
| Formula Weight |
945.2903 |
| InChI Formula |
InChI=1S/C44H51NO22/c1-20(46)56-18-33-35(60-22(3)48)37(61-23(4)49)40(64-26(7)52)43(66-33)67-36-34(19-57-21(2)47)65-42(39(63-25(6)51)38(36)62-24(5)50)58-17-32(41(53)54)45-44(55)59-16-31-29-14-10-8-12-27(29)28-13-9-11-15-30(28)31/h8-15,31-40,42-43H,16-19H2,1-7H3,(H,45,55)(H,53,54)/t32-,33+,34+,35-,36+,37-,38-,39+,40+,42+,43-/m0/s1 |
| IUPAC Name |
(2S)-3-[(2R,3R,4S,5R,6R)-3,4-diacetyloxy-6-(acetyloxymethyl)-5-[(2S,3R,4S,5S,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxyoxan-2-yl]oxy-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid |
| Molecular Formula |
C44H51NO22 |
| Molecular Weight (g/mol) |
945.87 |
| SMILES |
CC(=O)OC[C@H]1O[C@@H](OC[C@H](NC(=O)OCC2c3ccccc3c4ccccc24)C(=O)O)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1O[C@@H]5O[C@H](COC(=O)C)[C@H](OC(=O)C)[C@H](OC(=O)C)[C@H]5OC(=O)C |
| Synonym |
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-[2,3,6-tri-O-acetyl-4-O-(2,3,4,6-tetra-O-acetyl-beta-D-galactopyranosyl)-beta-D-glucopyranosyl]-L-serine, Hepta-O-acetyl-beta-D-lactosyl-Fmoc-Ser |
| Chemical Name or Material |
N-(9-Fluorenylmethoxycarbonyl)-O-(2,3,6,2',3',4',6'-hepta-O-acetyl-beta-D-lactosyl)-L-serine |