missing translation for 'onlineSavingsMsg'
Learn More

N-(7-Oxa-9,9,9-trifluorononyl)deoxynojirimycin, TRC

Product Code. 30424972
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30424972 10 mg
30419461 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30424972 Supplier Toronto Research Chemicals Supplier No. TRCO84515010MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 383417-50-3
Molecular Formula C14 H26 F3 N O5
Molecular Weight (g/mol) 345.36
IUPAC Name (2R,3R,4R,5S)-2-(hydroxymethyl)-1-[6-(2,2,2-trifluoroethoxy)hexyl]piperidine-3,4,5-triol
SMILES OC[C@@H]1[C@@H](O)[C@H](O)[C@@H](O)CN1CCCCCCOCC(F)(F)F

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H26F3NO5/c15-14(16,17)9-23-6-4-2-1-3-5-18-7-11(20)13(22)12(21)10(18)8-19/h10-13,19-22H,1-9H2/t10-,11+,12-,13-/m1/s1
Formula Weight 345.1763
Chemical Name or Material N-(7-Oxa-9,9,9-trifluorononyl)deoxynojirimycin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.