CAS RN 383417-50-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(7-Oxa-9,9,9-trifluorononyl)deoxynojirimycin, TRC
CAS: 383417-50-3 Molecular Formula: C14 H26 F3 N O5 Molecular Weight (g/mol): 345.36 IUPAC Name: (2R,3R,4R,5S)-2-(hydroxymethyl)-1-[6-(2,2,2-trifluoroethoxy)hexyl]piperidine-3,4,5-triol SMILES: OC[C@@H]1[C@@H](O)[C@H](O)[C@@H](O)CN1CCCCCCOCC(F)(F)F