missing translation for 'onlineSavingsMsg'
Learn More

N-(5-formylpyrimidin-2-yl)-N-methylglycine, TRC

Product Code. 30408081
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30408081 100 mg
30394434 50 mg
30390740 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408081 Supplier Toronto Research Chemicals Supplier No. TRCF691395100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 915921-77-6
Molecular Formula C8H9N3O3
Molecular Weight (g/mol) 195.17
SMILES CN(CC(=O)O)c1ncc(C=O)cn1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H9N3O3/c1-11(4-7(13)14)8-9-2-6(5-12)3-10-8/h2-3,5H,4H2,1H3,(H,13,14)
Formula Weight 195.0644
Chemical Name or Material N-(5-formylpyrimidin-2-yl)-N-methylglycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.