CAS RN 915921-77-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(5-formylpyrimidin-2-yl)-N-methylglycine, TRC
CAS: 915921-77-6 Molecular Formula: C8H9N3O3 Molecular Weight (g/mol): 195.17 SMILES: CN(CC(=O)O)c1ncc(C=O)cn1