missing translation for 'onlineSavingsMsg'
Learn More

N-[4-Hydroxy-3-(trifluoromethyl)phenyl]-3-methylurea, TRC

Product Code. 30389493
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389493 250 mg
30392505 500 mg
30407087 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389493 Supplier Toronto Research Chemicals Supplier No. TRCH645285250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1174758-89-4
Molecular Formula C9 H9 F3 N2 O2
Molecular Weight (g/mol) 234.18
Synonym Urea, N-[4-hydroxy-3-(trifluoromethyl)phenyl]-N'-methyl-, CGA 236432
IUPAC Name 1-[4-hydroxy-3-(trifluoromethyl)phenyl]-3-methylurea
SMILES CNC(=O)Nc1ccc(O)c(c1)C(F)(F)F

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H9F3N2O2/c1-13-8(16)14-5-2-3-7(15)6(4-5)9(10,11)12/h2-4,15H,1H3,(H2,13,14,16)
Formula Weight 234.0616
Chemical Name or Material Fluometuron-N-desmethyl-4-hydroxy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.