CAS RN 1174758-89-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-[4-Hydroxy-3-(trifluoromethyl)phenyl]-3-methylurea, TRC
CAS: 1174758-89-4 Molecular Formula: C9 H9 F3 N2 O2 Molecular Weight (g/mol): 234.18 Synonym: Urea, N-[4-hydroxy-3-(trifluoromethyl)phenyl]-N'-methyl-,CGA 236432 IUPAC Name: 1-[4-hydroxy-3-(trifluoromethyl)phenyl]-3-methylurea SMILES: CNC(=O)Nc1ccc(O)c(c1)C(F)(F)F